siRNA Details
| Gene Symbol: 1700110I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007160.1 | - | 1 | 62 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA464463.1 | - | 1 | 36 | Chr. 7 - 38408163 - 38442739 [+] on Build GRCm38 | |

1700110I07Rik
Genomic Map