siRNA Details
| Gene Symbol: 8030466E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033215.1 | - | 1 | 2327 | ||
| BC059088.1 | - | 1 | 405 | |||
| Gene Symbol: Trat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138735.1 | - | 4418 | |||

8030466E21Rik
Genomic Map