siRNA Details
Gene Symbol: C030034L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039918.1 | - | 666 | |||
AK081263.1 | - | 527 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728252.1 | - | 3 | 518 | Chr. 3 - 9403075 - 9435058 [+] on Build GRCm38 |
