siRNA Details
| Gene Symbol: C030034L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039918.1 | - | 1360 | |||
| AK081263.1 | - | 1221 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C030034L19Rik| Gene Symbol: C030034L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039918.1 | - | 1360 | |||
| AK081263.1 | - | 1221 | ||||