siRNA Details
| Gene Symbol: C030034L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039918.1 | - | 731 | |||
| AK081263.1 | - | 592 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728252.1 | - | 3 | 583 | Chr. 3 - 9403075 - 9435058 [+] on Build GRCm38 | |

C030034L19Rik
Genomic Map