siRNA Details
| Gene Symbol: A330105O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020736.1 | - | 1 | 97 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN531386.1 | - | 1 | 38 | Chr. 9 - 118579667 - 118580357 [+] on Build GRCm38 | |

A330105O20Rik
Genomic Map