siRNA Details
| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020223.1 | - | 1 | 468 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720465.1 | - | 1 | 468 | Chr. 1 - 86857371 - 86858005 [+] on Build GRCm38 | |

Dis3l2
Genomic Map