siRNA Details
| Gene Symbol: 8030488J09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020221.1 | - | 1 | 210 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720460.1 | - | 1 | 207 | Chr. 7 - 132575076 - 132575748 [-] on Build GRCm38 | |

8030488J09Rik
Genomic Map