siRNA Details
Gene Symbol: 8030488J09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020221.1 | - | 1 | 519 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720460.1 | - | 1 | 516 | Chr. 7 - 132575076 - 132575748 [-] on Build GRCm38 |
