siRNA Details
| Gene Symbol: Gm21221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077242.1 | 4 | 697 | |||
| GenBank mRNA | AK076945.1 | - | 4 | 700 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB017466.2 | - | 3 | 273 | Chr. 5 - 149801990 - 149804715 [-] on Build GRCm38 | |

Gm21221
Genomic Map