siRNA Details
| Gene Symbol: Gm30853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076946.1 | - | 6 | 759 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB017528.2 | - | 4 | 629 | Chr. 10 - 65875788 - 65898849 [-] on Build GRCm38 | |

Gm30853
Genomic Map