siRNA Details
| Gene Symbol: Gm16853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045742.1 | 2 | 309 | |||
| GenBank mRNA | AK076941.1 | - | 2 | 311 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615851.1 | - | 2 | 311 | Chr. 9 - 21406888 - 21411720 [-] on Build GRCm38 | |

Gm16853
Genomic Map