siRNA Details
| Gene Symbol: Gm16853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045742.1 | 4 | 980 | |||
| GenBank mRNA | AK076941.1 | - | 4 | 982 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16853| Gene Symbol: Gm16853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045742.1 | 4 | 980 | |||
| GenBank mRNA | AK076941.1 | - | 4 | 982 | ||