siRNA Details
| Gene Symbol: 4833417C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045187.1 | 2 | 744 | |||
| GenBank mRNA | AK014714.1 | - | 2 | 699 | ||
| AK133925.1 | - | 637 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714212.1 | - | 2 | 699 | Chr. 11 - 95858865 - 95860610 [+] on Build GRCm38 | |

4833417C18Rik
Genomic Map