siRNA Details
| Gene Symbol: 4833417C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045187.1 | 2 | 683 | |||
| GenBank mRNA | AK014714.1 | - | 2 | 638 | ||
| AK133925.1 | - | 576 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833417C18Rik| Gene Symbol: 4833417C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045187.1 | 2 | 683 | |||
| GenBank mRNA | AK014714.1 | - | 2 | 638 | ||
| AK133925.1 | - | 576 | ||||