siRNA Details
Gene Symbol: 4833417C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045187.1 | 1 | 147 | |||
GenBank mRNA | AK014714.1 | - | 1 | 102 | ||
AK133925.1 | - | 40 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4833417C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045187.1 | 1 | 147 | |||
GenBank mRNA | AK014714.1 | - | 1 | 102 | ||
AK133925.1 | - | 40 |