siRNA Details
| Gene Symbol: Snhg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036616.1 | - | 1 | 544 | ||
| AK051045.1 | - | 1 | 542 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | U40654.1 | - | 1 | 699 | Chr. - - [-] on Build | |

Snhg1
Genomic Map