siRNA Details
| Gene Symbol: Snhg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036616.1 | - | 1 | 620 | ||
| AK051045.1 | - | 1 | 618 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snhg1| Gene Symbol: Snhg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036616.1 | - | 1 | 620 | ||
| AK051045.1 | - | 1 | 618 | |||