siRNA Details
Gene Symbol: 4833418N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014719.1 | - | 2 | 766 | ||
AK028291.1 | - | 1 | 1918 | |||
AK082147.1 | - | 1 | 829 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4833418N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014719.1 | - | 2 | 766 | ||
AK028291.1 | - | 1 | 1918 | |||
AK082147.1 | - | 1 | 829 |