siRNA Details
| Gene Symbol: 4833418N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014719.1 | - | 2 | 625 | ||
| AK028291.1 | - | 1 | 1777 | |||
| AK082147.1 | - | 1 | 688 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833418N02Rik| Gene Symbol: 4833418N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014719.1 | - | 2 | 625 | ||
| AK028291.1 | - | 1 | 1777 | |||
| AK082147.1 | - | 1 | 688 | |||