siRNA Details
| Gene Symbol: Fam188b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030561.1 | - | 508 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB618028.1 | - | 2 | 508 | Chr. 3 - 58789903 - 58792019 [-] on Build GRCm38 | |

Fam188b2
Genomic Map