siRNA Details
| Gene Symbol: Fam188b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030561.1 | - | 2083 | |||
| BB026216.2 | - | 611 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam188b2| Gene Symbol: Fam188b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030561.1 | - | 2083 | |||
| BB026216.2 | - | 611 | ||||