siRNA Details
| Gene Symbol: Gm15558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134234.1 | - | 2 | 370 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA123006.1 | - | 2 | 398 | Chr. 16 - 10682384 - 10683569 [-] on Build GRCm38 | |

Gm15558
Genomic Map