siRNA Details
| Gene Symbol: Crkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003911.1 | - | 1 | 847 | ||
| AK087977.1 | - | 1 | 1772 | |||
| BY471344.1 | - | 1 | 355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crkl| Gene Symbol: Crkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003911.1 | - | 1 | 847 | ||
| AK087977.1 | - | 1 | 1772 | |||
| BY471344.1 | - | 1 | 355 | |||