siRNA Details
| Gene Symbol: 2900003A17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013479.1 | - | 1 | 411 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712445.1 | - | 1 | 411 | Chr. 12 - 25061784 - 25062314 [+] on Build GRCm38 | |

2900003A17Rik
Genomic Map