siRNA Details
| Gene Symbol: 4930478P22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046060.1 | 2, 3 | 1198 | |||
| GenBank mRNA | AK015590.1 | - | 2, 3 | 759 | ||
| AK016658.1 | - | 2, 3 | 1199 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930478P22Rik| Gene Symbol: 4930478P22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046060.1 | 2, 3 | 1198 | |||
| GenBank mRNA | AK015590.1 | - | 2, 3 | 759 | ||
| AK016658.1 | - | 2, 3 | 1199 | |||