siRNA Details
| Gene Symbol: 4930478P22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046060.1 | 1 | 537 | |||
| GenBank mRNA | AK015590.1 | - | 1 | 550 | ||
| AK016658.1 | - | 1 | 538 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715140.1 | - | 1 | 551 | Chr. 5 - 35361541 - 35367030 [+] on Build GRCm38 | |

4930478P22Rik
Genomic Map