siRNA Details
Gene Symbol: 4930478P22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046060.1 | 3, 4 | 1231 | |||
GenBank mRNA | AK015590.1 | - | 3, 4 | 792 | ||
AK016658.1 | - | 3, 4 | 1232 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930478P22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046060.1 | 3, 4 | 1231 | |||
GenBank mRNA | AK015590.1 | - | 3, 4 | 792 | ||
AK016658.1 | - | 3, 4 | 1232 |