siRNA Details
Gene Symbol: 4930479D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046277.1 | 2 | 511 | |||
GenBank mRNA | AK015591.1 | - | 2 | 512 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930479D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046277.1 | 2 | 511 | |||
GenBank mRNA | AK015591.1 | - | 2 | 512 |