siRNA Details
| Gene Symbol: 4930402F11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045940.1 | 3 | 505 | |||
| GenBank mRNA | AK015048.1 | - | 3 | 507 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930402F11Rik| Gene Symbol: 4930402F11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045940.1 | 3 | 505 | |||
| GenBank mRNA | AK015048.1 | - | 3 | 507 | ||