siRNA Details
| Gene Symbol: 2700005E23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA146492.1 | - | 1 | 329 | ||
| AK012210.1 | - | 1 | 92 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2700005E23Rik| Gene Symbol: 2700005E23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA146492.1 | - | 1 | 329 | ||
| AK012210.1 | - | 1 | 92 | |||