siRNA Details
| Gene Symbol: 2700005E23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012210.1 | - | 1 | 278 | ||
| AV123326.1 | - | 1 | 166 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711388.1 | - | 1 | 278 | Chr. 14 - 118091370 - 118091777 [+] on Build GRCm38 | |

2700005E23Rik
Genomic Map