siRNA Details
| Gene Symbol: Cfap54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015623.1 | - | 1 | 59 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715174.1 | - | 1 | 60 | Chr. 10 - 93081306 - 93081902 [+] on Build GRCm38 | |

Cfap54
Genomic Map