siRNA Details
| Gene Symbol: Zim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007312.2 | - | 502 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707418.1 | - | 3 | 502 | Chr. 7 - 6976302 - 6977420 [-] on Build GRCm38 | |

Zim3
Genomic Map