siRNA Details
| Gene Symbol: Gm11730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086245.1 | - | 3 | 624 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB523703.2 | - | 507 | Chr. - - [-] on Build | ||

Gm11730
Genomic Map