siRNA Details
| Gene Symbol: Gm11730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086245.1 | - | 1 | 255 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB662708.1 | - | 1 | 255 | Chr. 11 - 117095639 - 117097029 [-] on Build GRCm38 | |

Gm11730
Genomic Map