siRNA Details
| Gene Symbol: Gm19784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040461.1 | 5 | 787 | |||
| GenBank mRNA | AK136089.1 | - | 2 | 435 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ052949.1 | - | 2 | 435 | Chr. 18 - 66925143 - 66926373 [-] on Build GRCm38 | |

Gm19784
Genomic Map