siRNA Details
| Gene Symbol: 4930568A12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016244.1 | - | 1 | 29 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715855.1 | - | 27 | Chr. - - [-] on Build | ||

4930568A12Rik
Genomic Map