siRNA Details
| Gene Symbol: 5033425G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017193.1 | - | 1244 | |||
| BB323924.2 | - | 666 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5033425G24Rik| Gene Symbol: 5033425G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017193.1 | - | 1244 | |||
| BB323924.2 | - | 666 | ||||