siRNA Details
| Gene Symbol: 5033424D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017190.1 | - | 1 | 434 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716920.1 | - | 1 | 430 | Chr. 12 - 108878772 - 108879418 [+] on Build GRCm38 | |

5033424D13Rik
Genomic Map