siRNA Details
Gene Symbol: LOC102631757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110502.1 | 3 | 485 | |||
GenBank mRNA | AK085535.1 | - | 3 | 486 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102631757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110502.1 | 3 | 485 | |||
GenBank mRNA | AK085535.1 | - | 3 | 486 |