siRNA Details
| Gene Symbol: LOC102631757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110502.1 | 6 | 736 | |||
| NR_110503.1 | 6 | 632 | ||||
| GenBank mRNA | AK050258.1 | - | 6 | 590 | ||
| AK085535.1 | - | 6 | 737 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB502343.2 | - | 5 | 457 | Chr. 17 - 31153928 - 31162323 [+] on Build GRCm38 | |

LOC102631757
Genomic Map