siRNA Details
| Gene Symbol: LOC102631757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085535.1 | - | 1 | 83 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB661409.1 | - | 1 | 83 | Chr. 17 - 31152752 - 31161441 [+] on Build GRCm38 | |

LOC102631757
Genomic Map