siRNA Details
| Gene Symbol: Gm16796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040367.1 | 2, 3 | 2765 | |||
| GenBank mRNA | AK138533.1 | - | 2, 3 | 2766 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16796| Gene Symbol: Gm16796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040367.1 | 2, 3 | 2765 | |||
| GenBank mRNA | AK138533.1 | - | 2, 3 | 2766 | ||