siRNA Details
| Gene Symbol: 4930543I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016029.1 | - | 3 | 337 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715609.1 | - | 3 | 337 | Chr. 15 - 86737826 - 86766127 [+] on Build GRCm38 | |

4930543I03Rik
Genomic Map