siRNA Details
| Gene Symbol: 4933407E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045819.1 | 3 | 472 | |||
| GenBank mRNA | AK160260.1 | - | 3 | 461 | ||
| BB616969.1 | - | 3 | 461 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615438.1 | - | 3 | 457 | Chr. 4 - 124569195 - 124574414 [+] on Build GRCm38 | |

4933407E24Rik
Genomic Map