siRNA Details
| Gene Symbol: 4930483J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049560.1 | - | 1 | 128 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA464812.1 | - | 1 | 131 | Chr. 15 - 81190476 - 81192392 [+] on Build GRCm38 | |

4930483J18Rik
Genomic Map