siRNA Details
| Gene Symbol: Gm15713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046026.1 | 2 | 147 | |||
| GenBank mRNA | AK042323.1 | - | 2 | 148 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB639217.1 | - | 2 | 148 | Chr. 16 - 43411626 - 43420196 [-] on Build GRCm38 | |

Gm15713
Genomic Map