siRNA Details
| Gene Symbol: 9430081H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020500.1 | - | 1 | 103 | ||
| AK040656.1 | - | 1 | 257 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721875.1 | - | 1 | 103 | Chr. 9 - 44378741 - 44379296 [-] on Build GRCm38 | |

9430081H08Rik
Genomic Map