siRNA Details
| Gene Symbol: Gemin4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028076.1 | - | 2 | 69 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706277.1 | - | 2 | 69 | Chr. 11 - 76217079 - 76217515 [-] on Build GRCm38 | |

Gemin4
Genomic Map